Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PMCH
Entrez Gene ID
5367
Gene Name
pro-melanin concentrating hormone
Gene Aliases
MCH, ppMCH
Chromosome Location
Chr.12: 102196459 - 102199540 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002674.3 NP_002665.2 2 - 3 522 116
GenBank mRNA AV723033.1 - - 212 116
BC018048.1 - 2 - 3 510 116
CB047052.1 - 1 - 2 116
M57703.1 - 2 - 3 501 116

View Full Product Details