Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CILP
Entrez Gene ID
8483
Gene Name
cartilage intermediate layer protein
Gene Aliases
CILP-1, HsT18872
Chromosome Location
Chr.15: 65195999 - 65211608 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003613.3 NP_003604.3 2 - 3 243 61
GenBank mRNA AF035408.1 - 2 - 3 191 61
AK225652.1 - 2 - 3 210 61
AK225926.1 - 2 - 3 210 61
AK313352.1 - 2 - 3 214 61
AL602624.1 - 2 - 3 214 61
AY358904.1 - 2 - 3 197 61
BC035776.1 - 2 - 3 190 61
DB071945.1 - 2 - 3 245 61

View Full Product Details