Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- DNASE1
- Entrez Gene ID
-
1773
- Gene Name
- deoxyribonuclease I
- Gene Aliases
-
DNL1, DRNI
- Chromosome Location
-
Chr.16: 3611705 - 3665472 on Build GRCh38
View Full Product Details