The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005538.3 | NP_005529.1 | 1 - 2 | 515 | 70 |
GenBank mRNA | AK314849.1 | - | 1 - 2 | 487 | 70 |
BC020693.1 | - | 1 - 2 | 457 | 70 | |
BC130324.1 | - | 1 - 2 | 396 | 70 | |
BC130326.1 | - | 1 - 2 | 396 | 70 | |
BP237373.1 | - | 1 - 2 | 515 | 70 |