Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CALCRL
Entrez Gene ID
10203
Gene Name
calcitonin receptor like receptor
Gene Aliases
CGRPR, CRLR
Chromosome Location
Chr.2: 187341963 - 187448307 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271751.1 NP_001258680.1 13 - 14 1557 130
NM_005795.5 NP_005786.1 14 - 15 1722 130
XM_005246231.3 XP_005246288.2 15 - 16 1766 130
XM_005246232.3 XP_005246289.2 16 - 17 1807 130
XM_005246234.3 XP_005246291.2 14 - 15 1638 130
XM_017003151.1 XP_016858640.1 15 - 16 1697 130
GenBank mRNA AK291630.1 - 14 - 15 1680 130
AK292998.1 - 13 - 14 1515 130
AY389506.1 - 11 - 12 1174 130
CB270158.1 - 2 - 3 115 130
L76380.1 - 14 - 15 1728 130
U17473.1 - 14 - 15 1644 130

View Full Product Details