The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001200053.1 | NP_001186982.1 | 8 - 9 | 1640 | 86 |
NM_001200054.1 | NP_001186983.1 | 10 - 11 | 1919 | 86 | |
NM_001320121.1 | NP_001307050.1 | 11 - 12 | 1793 | 86 | |
NM_001320122.1 | NP_001307051.1 | 11 - 12 | 1772 | 86 | |
NM_006928.4 | NP_008859.1 | 11 - 12 | 1933 | 86 | |
XM_006719569.1 | XP_006719632.1 | 10 - 11 | 2510 | 86 | |
XM_011538685.1 | XP_011536987.1 | 11 - 12 | 1983 | 86 | |
GenBank mRNA | AK092881.1 | - | 10 - 11 | 2511 | 86 |
AK130180.1 | - | 11 - 12 | 1957 | 86 | |
AK300150.1 | - | 8 - 9 | 1620 | 86 | |
BC001414.2 | - | 10 - 11 | 1857 | 86 | |
BT007202.1 | - | 10 - 11 | 1847 | 86 | |
CA432578.1 | - | 1 - 2 | 86 | ||
M32295.1 | - | 10 - 11 | 1868 | 86 | |
M77348.1 | - | 10 - 11 | 1879 | 86 | |
S73003.1 | - | 10 - 11 | 1868 | 86 | |
U01874.1 | - | 10 - 11 | 1856 | 86 |