The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000572.2 | NP_000563.1 | 3 - 4 | 441 | 119 |
GenBank mRNA | AF043333.1 | - | 3 - 4 | 328 | 119 |
AY029171.1 | - | 3 - 4 | 382 | 119 | |
BC104252.1 | - | 3 - 4 | 438 | 119 | |
BC104253.1 | - | 3 - 4 | 438 | 119 | |
CR541993.1 | - | 3 - 4 | 382 | 119 | |
M57627.1 | - | 3 - 4 | 412 | 119 |