The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001123041.2 | NP_001116513.2 | 2 - 3 | 1424 | 74 |
GenBank mRNA | BC074751.2 | - | 1 - 2 | 978 | 74 |
BC126452.1 | - | 1 - 2 | 973 | 74 | |
BC136396.1 | - | 1 - 2 | 973 | 74 | |
U03882.1 | - | 1 - 2 | 978 | 74 |