Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITGB5
Entrez Gene ID
3693
Gene Name
integrin subunit beta 5
Gene Aliases
-
Chromosome Location
Chr.3: 124761948 - 124901411 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002213.4 NP_002204.2 11 - 12 2221 78
XM_005247436.2 XP_005247493.1 10 - 11 2364 78
XM_006713630.2 XP_006713693.1 10 - 11 2319 78
XM_017006351.1 XP_016861840.1 11 - 12 1992 78
XM_017006352.1 XP_016861841.1 11 - 12 1931 78
XM_017006353.1 XP_016861842.1 11 - 12 1963 78
XM_017006354.1 XP_016861843.1 8 - 9 2106 78
XM_017006355.1 XP_016861844.1 10 - 11 1841 78
GenBank mRNA AK309551.1 - 3 - 4 707 78
AK315429.1 - 11 - 12 1957 78
BC006541.2 - 11 - 12 2213 78
J05633.1 - 11 - 12 1962 78
M35011.1 - 11 - 12 1945 78
X53002.1 - 11 - 12 2252 78

View Full Product Details