Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDE4D
Entrez Gene ID
5144
Gene Name
phosphodiesterase 4D
Gene Aliases
ACRDYS2, DPDE3, HSPDE4D, PDE43, PDE4DN2, STRK1
Chromosome Location
Chr.5: 58969038 - 60488098 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104631.1 NP_001098101.1 3 - 4 856 70
NM_001165899.1 NP_001159371.1 5 - 6 747 70
NM_001197218.1 NP_001184147.1 3 - 4 595 70
NM_001197219.1 NP_001184148.1 3 - 4 373 70
NM_001197220.1 NP_001184149.1 3 - 4 1011 70
NM_006203.4 NP_006194.2 3 - 4 399 70
XM_005248537.3 XP_005248594.1 3 - 4 1553 70
XM_005248538.4 XP_005248595.1 4 - 5 3454 70
XM_011543469.1 XP_011541771.1 6 - 7 836 70
XM_011543470.2 XP_011541772.1 6 - 7 760 70
XM_011543471.1 XP_011541773.1 5 - 6 713 70
XM_011543473.1 XP_011541775.1 5 - 6 661 70
XM_011543477.2 XP_011541779.1 5 - 6 663 70
XM_017009565.1 XP_016865054.1 6 - 7 873 70
XM_017009566.1 XP_016865055.1 6 - 7 763 70
XM_017009567.1 XP_016865056.1 4 - 5 501 70
XM_017009568.1 XP_016865057.1 6 - 7 747 70
XM_017009569.1 XP_016865058.1 5 - 6 626 70
GenBank mRNA AF012073.1 - 3 - 4 609 70
AF536976.1 - 4 - 5 570 70
AF536977.1 - 3 - 4 373 70
AJ250854.1 - 3 - 4 492 70
AJ250855.1 - 3 - 4 347 70
AY245866.1 - 5 - 6 712 70
AY245867.1 - 3 - 4 1011 70
AY388960.1 - 3 - 4 294 70
BC008390.1 - 3 - 4 619 70
BT007398.1 - 3 - 4 492 70
L20970.1 - 3 - 4 407 70
U02882.1 - 2 - 3 177 70

View Full Product Details