The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001104631.1 | NP_001098101.1 | 8 - 9 | 1358 | 123 |
NM_001165899.1 | NP_001159371.1 | 10 - 11 | 1249 | 123 | |
NM_001197218.1 | NP_001184147.1 | 8 - 9 | 1097 | 123 | |
NM_001197219.1 | NP_001184148.1 | 8 - 9 | 875 | 123 | |
NM_001197220.1 | NP_001184149.1 | 8 - 9 | 1513 | 123 | |
NM_001197221.1 | NP_001184150.1 | 4 - 5 | 833 | 123 | |
NM_001197222.1 | NP_001184151.1 | 3 - 4 | 919 | 123 | |
NM_001197223.1 | NP_001184152.1 | 3 - 4 | 709 | 123 | |
NM_006203.4 | NP_006194.2 | 8 - 9 | 901 | 123 | |
XM_005248537.3 | XP_005248594.1 | 8 - 9 | 2055 | 123 | |
XM_005248538.4 | XP_005248595.1 | 9 - 10 | 3956 | 123 | |
XM_011543469.1 | XP_011541771.1 | 11 - 12 | 1338 | 123 | |
XM_011543470.2 | XP_011541772.1 | 11 - 12 | 1262 | 123 | |
XM_011543471.1 | XP_011541773.1 | 10 - 11 | 1215 | 123 | |
XM_011543473.1 | XP_011541775.1 | 10 - 11 | 1163 | 123 | |
XM_011543477.2 | XP_011541779.1 | 10 - 11 | 1165 | 123 | |
XM_017009565.1 | XP_016865054.1 | 11 - 12 | 1375 | 123 | |
XM_017009566.1 | XP_016865055.1 | 11 - 12 | 1265 | 123 | |
XM_017009567.1 | XP_016865056.1 | 9 - 10 | 1003 | 123 | |
XM_017009568.1 | XP_016865057.1 | 11 - 12 | 1249 | 123 | |
XM_017009569.1 | XP_016865058.1 | 10 - 11 | 1128 | 123 | |
XM_017009570.1 | XP_016865059.1 | 7 - 8 | 1948 | 123 | |
XM_017009571.1 | XP_016865060.1 | 10 - 11 | 1171 | 123 | |
GenBank mRNA | AF012073.1 | - | 8 - 9 | 1111 | 123 |
AF012074.1 | - | 4 - 5 | 847 | 123 | |
AF536975.1 | - | 3 - 4 | 516 | 123 | |
AF536976.1 | - | 10 - 11 | 1072 | 123 | |
AF536977.1 | - | 8 - 9 | 875 | 123 | |
AJ250852.1 | - | 4 - 5 | 443 | 123 | |
AJ250854.1 | - | 9 - 10 | 1038 | 123 | |
AY245866.1 | - | 10 - 11 | 1214 | 123 | |
AY245867.1 | - | 8 - 9 | 1513 | 123 | |
AY388960.1 | - | 8 - 9 | 796 | 123 | |
BC036319.1 | - | 3 - 4 | 711 | 123 | |
L20970.1 | - | 8 - 9 | 909 | 123 | |
U02882.1 | - | 7 - 8 | 679 | 123 | |
U50157.1 | - | 3 - 4 | 517 | 123 | |
U50158.1 | - | 4 - 5 | 431 | 123 | |
U50159.1 | - | 8 - 9 | 852 | 123 | |
U79571.1 | - | 3 - 4 | 922 | 123 |