Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSHB
Entrez Gene ID
7252
Gene Name
thyroid stimulating hormone beta
Gene Aliases
TSH-B, TSH-BETA
Chromosome Location
Chr.1: 115029824 - 115035935 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000549.4 NP_000540.2 2 - 3 196 72
XM_011542065.2 XP_011540367.1 1 - 2 235 72
GenBank mRNA BC069298.1 - 1 - 2 158 72

View Full Product Details