The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135690.2 | NP_001129162.1 | 3 - 4 | 364 | 56 |
GenBank mRNA | AK314908.1 | - | 3 - 4 | 305 | 56 |
BC032505.1 | - | 1 - 2 | 215 | 56 | |
BI913625.1 | - | 1 - 2 | 219 | 56 | |
BP370091.1 | - | 3 - 4 | 366 | 56 | |
CR541808.1 | - | 1 - 2 | 139 | 56 | |
CR541828.1 | - | 1 - 2 | 139 | 56 | |
DB496514.1 | - | 3 - 4 | 354 | 56 |