Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DPP4
Entrez Gene ID
1803
Gene Name
dipeptidyl peptidase 4
Gene Aliases
ADABP, ADCP2, CD26, DPPIV, TP103
Chromosome Location
Chr.2: 161992241 - 162074542 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001935.3 NP_001926.2 5 - 6 928 115
XM_005246371.2 XP_005246428.1 5 - 6 505 115
GenBank mRNA AB451339.1 - 4 - 5 367 115
AB451488.1 - 4 - 5 367 115
AK314798.1 - 5 - 6 505 115
AY429532.1 - 5 - 6 462 115
BC013329.2 - 4 - 5 362 115
BC065265.1 - 5 - 6 928 115
M74777.1 - 5 - 6 377 115
M80536.1 - 4 - 5 367 115
X60708.1 - 5 - 6 442 115

View Full Product Details