Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TCF7
Entrez Gene ID
6932
Gene Name
transcription factor 7 (T-cell specific, HMG-box)
Gene Aliases
TCF-1
Chromosome Location
Chr.5: 134114703 - 134148229 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134851.2 NP_001128323.2 4 - 5 392 105
NM_003202.3 NP_003193.2 5 - 6 836 105
NM_201632.3 NP_963963.1 4 - 5 392 105
NM_201634.3 NP_963965.1 4 - 5 392 105
NM_213648.3 NP_998813.1 3 - 4 626 105
NR_033449.1 3 - 4 627 105
XM_006714679.3 XP_006714742.1 5 - 6 844 105
XM_006714684.2 XP_006714747.1 5 - 6 844 105
XM_006714686.3 XP_006714749.1 4 - 5 391 105
XM_011543609.2 XP_011541911.1 3 - 4 338 105
XM_011543612.2 XP_011541914.1 3 - 4 338 105
XM_011543613.2 XP_011541915.1 3 - 4 377 105
GenBank mRNA AK057580.1 - 3 - 4 627 105
AK093530.1 - 3 - 4 497 105
AK310591.1 - 3 - 4 338 105
AL832816.1 - 5 - 6 552 105
BC048769.1 - 5 - 6 836 105
BC072023.1 - 5 - 6 771 105
DA627229.1 - 4 - 5 392 105
DB145426.1 - 4 - 5 392 105
X59869.1 - 4 - 5 374 105
X59870.1 - 4 - 5 374 105
X59871.1 - 4 - 5 374 105
Z47361.1 - 3 - 4 296 105
Z47362.1 - 3 - 4 296 105

View Full Product Details