The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000154.1 | NP_000145.1 | 5 - 6 | 858 | 64 |
GenBank mRNA | AK130064.1 | - | - | 1148 | 64 |
AK303832.1 | - | 5 - 6 | 895 | 64 | |
AK314890.1 | - | 5 - 6 | 818 | 64 | |
BC001166.1 | - | 5 - 6 | 839 | 64 | |
BT007005.1 | - | 5 - 6 | 795 | 64 | |
EU794610.1 | - | 5 - 6 | 823 | 64 | |
U26401.1 | - | 5 - 6 | 823 | 64 |