The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000455.4 | NP_000446.1 | 8 - 9 | 2224 | 92 |
GenBank mRNA | AB209553.1 | - | 7 - 8 | 1807 | 92 |
AF035625.1 | - | 8 - 9 | 1447 | 92 | |
AF217978.1 | - | 9 - 10 | 2070 | 92 | |
AK314858.1 | - | 8 - 9 | 1235 | 92 | |
BC007981.2 | - | 8 - 9 | 1559 | 92 | |
BC019334.1 | - | 8 - 9 | 1380 | 92 | |
EF644172.1 | - | 8 - 9 | 1193 | 92 | |
KU178339.1 | - | 8 - 9 | 1109 | 92 | |
U63333.1 | - | 8 - 9 | 1109 | 92 |