The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204.6 | NP_001195.2 | 1 - 2 | 1226 | 69 |
XM_011511687.1 | XP_011509989.1 | 1 - 2 | 1243 | 69 | |
GenBank mRNA | AK292430.1 | - | 1 - 2 | 490 | 69 |
BC052985.2 | - | 1 - 2 | 652 | 69 | |
D50516.1 | - | 1 - 2 | 486 | 69 | |
U20165.1 | - | 1 - 2 | 114 | 69 | |
Z48923.1 | - | 1 - 2 | 459 | 69 |