The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165038.1 | NP_001158510.1 | 5 - 6 | 1410 | 88 |
NM_001165039.1 | NP_001158511.1 | 4 - 5 | 1326 | 88 | |
NM_001495.4 | NP_001486.4 | 6 - 7 | 1725 | 88 | |
XM_006716327.3 | XP_006716390.1 | 7 - 8 | 1171 | 88 | |
XM_011544484.2 | XP_011542786.1 | 7 - 8 | 1203 | 88 | |
GenBank mRNA | AF002700.1 | - | 6 - 7 | 1085 | 88 |
AK299623.1 | - | 5 - 6 | 816 | 88 | |
AY326396.1 | - | 4 - 5 | 651 | 88 | |
AY941828.1 | - | 5 - 6 | 735 | 88 | |
BC041688.1 | - | 6 - 7 | 1725 | 88 | |
U93703.1 | - | 6 - 7 | 1050 | 88 | |
U97145.1 | - | 6 - 7 | 1303 | 88 |