Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 40300762-40301250 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL9A2
Entrez Gene ID
1298
Gene Name
collagen type IX alpha 2
Gene Aliases
DJ39G22.4, EDM2, MED, STL5
UniGene
Hs.418012
Gene Chromosome Location
Chr.1: 40300487-40317653 on Build GRCh38
Transcripts
NM_001852.3 XM_006710365.3 XM_011540715.2 XM_011540716.2 XM_011540717.2 XM_017000332.1 XM_017000333.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3489540 c.2020G>A p.A674T Chr.1: 40301232-40301232 on Build GRCh38
290046 c.2036G>C p.R679P Chr.1: 40301216-40301216 on Build GRCh38
6852961 c.2002G>A p.A668T Chr.1: 40301250-40301250 on Build GRCh38
1342615 c.2036G>A p.R679H Chr.1: 40301216-40301216 on Build GRCh38
414922 c.2012G>C p.G671A Chr.1: 40301240-40301240 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs558294493 Chr.1: 40300828-40300828 on Build GRCh38
rs969999104 Chr.1: 40301033-40301033 on Build GRCh38
rs547105794 Chr.1: 40301144-40301144 on Build GRCh38
rs553963794 Chr.1: 40301115-40301115 on Build GRCh38
rs1036640585 Chr.1: 40301064-40301064 on Build GRCh38
rs968288876 Chr.1: 40301228-40301228 on Build GRCh38
rs538062384 Chr.1: 40300819-40300819 on Build GRCh38
rs994507222 Chr.1: 40300886-40300886 on Build GRCh38
rs187788913 Chr.1: 40300919-40300919 on Build GRCh38
rs898091801 Chr.1: 40301005-40301005 on Build GRCh38
rs749873890 Chr.1: 40301202-40301202 on Build GRCh38
rs775823584 Chr.1: 40301176-40301176 on Build GRCh38
rs2747510 Chr.1: 40300868-40300868 on Build GRCh38
rs765273618 Chr.1: 40301201-40301201 on Build GRCh38
rs148008235 Chr.1: 40301233-40301233 on Build GRCh38
rs561657830 Chr.1: 40300960-40300960 on Build GRCh38
rs749753328 Chr.1: 40301140-40301140 on Build GRCh38
rs545871447 Chr.1: 40300915-40300915 on Build GRCh38
rs746019124 Chr.1: 40301163-40301163 on Build GRCh38
rs973618316 Chr.1: 40300804-40300804 on Build GRCh38
rs534465543 Chr.1: 40301082-40301082 on Build GRCh38
rs954102697 Chr.1: 40301016-40301016 on Build GRCh38
rs115675008 Chr.1: 40301194-40301194 on Build GRCh38
rs913822150 Chr.1: 40300912-40300912 on Build GRCh38
rs548912125 Chr.1: 40300812-40300812 on Build GRCh38
rs201847956 Chr.1: 40301193-40301193 on Build GRCh38
rs146173817 Chr.1: 40300931-40300931 on Build GRCh38
rs980874762 Chr.1: 40300822-40300822 on Build GRCh38
rs114634369 Chr.1: 40301071-40301071 on Build GRCh38
rs1027333704 Chr.1: 40300901-40300901 on Build GRCh38
rs866372518 Chr.1: 40300838-40300838 on Build GRCh38
rs749736976 Chr.1: 40301232-40301232 on Build GRCh38
rs962569332 Chr.1: 40300795-40300795 on Build GRCh38
rs1057515524 Chr.1: 40300796-40300796 on Build GRCh38
rs912674043 Chr.1: 40301059-40301059 on Build GRCh38
rs778302764 Chr.1: 40301134-40301134 on Build GRCh38
rs930560318 Chr.1: 40301112-40301112 on Build GRCh38
rs904873014 Chr.1: 40301157-40301157 on Build GRCh38
rs781727058 Chr.1: 40301144-40301145 on Build GRCh38
rs773649280 Chr.1: 40300813-40300813 on Build GRCh38
rs199897562 Chr.1: 40301216-40301216 on Build GRCh38
rs968059509 Chr.1: 40301089-40301089 on Build GRCh38
rs932745561 Chr.1: 40300796-40300796 on Build GRCh38
rs1057515525 Chr.1: 40300952-40300952 on Build GRCh38
rs1036771449 Chr.1: 40301219-40301219 on Build GRCh38
rs778216645 Chr.1: 40301230-40301230 on Build GRCh38
rs921340380 Chr.1: 40300785-40300785 on Build GRCh38
rs760921400 Chr.1: 40301186-40301186 on Build GRCh38
rs986858139 Chr.1: 40301031-40301031 on Build GRCh38
rs764864718 Chr.1: 40301172-40301172 on Build GRCh38
rs761675654 Chr.1: 40301198-40301198 on Build GRCh38
rs564283453 Chr.1: 40301070-40301070 on Build GRCh38
rs913862708 Chr.1: 40301083-40301083 on Build GRCh38
rs928030014 Chr.1: 40300841-40300841 on Build GRCh38
rs979421855 Chr.1: 40300947-40300947 on Build GRCh38
rs1057515566 Chr.1: 40300860-40300860 on Build GRCh38
rs2229827 Chr.1: 40301060-40301060 on Build GRCh38
rs754688437 Chr.1: 40301223-40301223 on Build GRCh38
rs35948159 Chr.1: 40301121-40301121 on Build GRCh38
rs41480445 Chr.1: 40300946-40300946 on Build GRCh38
rs995523986 Chr.1: 40301015-40301015 on Build GRCh38
rs890843821 Chr.1: 40300937-40300940 on Build GRCh38
rs1021202446 Chr.1: 40300936-40300936 on Build GRCh38
rs1015192473 Chr.1: 40300952-40300952 on Build GRCh38
rs921264993 Chr.1: 40300984-40300984 on Build GRCh38
rs944591500 Chr.1: 40300852-40300852 on Build GRCh38
rs199547057 Chr.1: 40301217-40301217 on Build GRCh38
rs774289715 Chr.1: 40301145-40301145 on Build GRCh38
rs980925793 Chr.1: 40301058-40301058 on Build GRCh38
rs1001972451 Chr.1: 40300909-40300909 on Build GRCh38
rs774784075 Chr.1: 40300905-40300905 on Build GRCh38
rs1036267607 Chr.1: 40300856-40300856 on Build GRCh38
rs41466547 Chr.1: 40300870-40300870 on Build GRCh38
rs137890492 Chr.1: 40300981-40300981 on Build GRCh38
rs757963762 Chr.1: 40301215-40301215 on Build GRCh38
rs771268496 Chr.1: 40300904-40300904 on Build GRCh38

View Full Product Details