Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HCK
Entrez Gene ID
3055
Gene Name
HCK proto-oncogene, Src family tyrosine kinase
Gene Aliases
JTK9, p59Hck, p61Hck
Chromosome Location
Chr.20: 32052188 - 32101856 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172129.1 NP_001165600.1 6 - 7 765 64
NM_001172130.1 NP_001165601.1 6 - 7 762 64
NM_001172131.1 NP_001165602.1 6 - 7 762 64
NM_001172132.1 NP_001165603.1 7 - 8 831 64
NM_001172133.1 NP_001165604.1 7 - 8 829 64
NM_002110.3 NP_002101.2 6 - 7 765 64
GenBank mRNA AK026432.1 - 6 - 7 699 64
AK225819.1 - 6 - 7 711 64
AK290928.1 - 7 - 8 777 64
AK298726.1 - 7 - 8 775 64
AK314335.1 - 6 - 7 678 64
BC014435.1 - 6 - 7 651 64
BC094847.1 - 6 - 7 662 64
BC108930.1 - 6 - 7 692 64
BC108931.1 - 6 - 7 692 64
BC113854.1 - 6 - 7 689 64
BC114463.1 - 6 - 7 692 64
M16592.1 - 5 - 6 540 64

View Full Product Details