The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003328.2 | NP_003319.2 | 11 - 12 | 1265 | 113 |
XM_011513747.2 | XP_011512049.1 | 10 - 11 | 1217 | 113 | |
XM_011513748.2 | XP_011512050.1 | 4 - 5 | 553 | 113 | |
XM_017008580.1 | XP_016864069.1 | 12 - 13 | 2467 | 113 | |
XM_017008581.1 | XP_016864070.1 | 10 - 11 | 2276 | 113 | |
XM_017008582.1 | XP_016864071.1 | 5 - 6 | 700 | 113 | |
GenBank mRNA | AK300141.1 | - | 1 - 2 | 275 | 113 |
L27071.1 | - | 11 - 12 | 1265 | 113 |