Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SQSTM1
Entrez Gene ID
8878
Gene Name
sequestosome 1
Gene Aliases
A170, FTDALS3, OSIL, PDB3, ZIP3, p60, p62, p62B
Chromosome Location
Chr.5: 179806388 - 179838078 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142298.1 NP_001135770.1 6 - 7 863 55
NM_001142299.1 NP_001135771.1 6 - 7 780 55
NM_003900.4 NP_003891.1 5 - 6 850 55
XM_017010010.1 XP_016865499.1 5 - 6 1082 55
GenBank mRNA AK025146.1 - 5 - 6 569 55
AK098077.1 - 4 - 5 1023 55
AK226167.1 - 5 - 6 572 55
AK293509.1 - 5 - 6 740 55
AK304877.1 - 6 - 7 629 55
AK312451.1 - 5 - 6 814 55
BC000951.2 - 4 - 5 539 55
BC001874.1 - 7 - 8 1014 55
BC003139.1 - 5 - 6 773 55
BC005857.2 - 4 - 5 513 55
BC017222.1 - 5 - 6 795 55
BC019111.1 - 5 - 6 787 55
U41806.1 - 5 - 6 775 55
U46751.1 - 5 - 6 807 55

View Full Product Details