The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010938.1 | NP_001010938.1 | 14 - 15 | 3402 | 100 |
NM_001308046.1 | NP_001294975.1 | 13 - 14 | 3415 | 100 | |
NM_005781.4 | NP_005772.3 | 14 - 15 | 3656 | 100 | |
XM_005269268.3 | XP_005269325.1 | 15 - 16 | 3498 | 100 | |
XM_005269270.3 | XP_005269327.1 | 15 - 16 | 3346 | 100 | |
XM_005269274.3 | XP_005269331.1 | 12 - 13 | 2763 | 100 | |
XM_005269275.3 | XP_005269332.1 | 14 - 15 | 3242 | 100 | |
XM_011512317.2 | XP_011510619.1 | 17 - 18 | 4065 | 100 | |
XM_011512318.2 | XP_011510620.2 | 15 - 16 | 3431 | 100 | |
XM_011512320.1 | XP_011510622.1 | 15 - 16 | 3220 | 100 | |
XM_011512321.2 | XP_011510623.1 | 13 - 14 | 3037 | 100 | |
XM_017005508.1 | XP_016860997.1 | 14 - 15 | 3392 | 100 | |
XM_017005510.1 | XP_016860999.1 | 14 - 15 | 3341 | 100 | |
GenBank mRNA | AK126712.1 | - | 3 - 4 | 2559 | 100 |
AK127455.1 | - | 6 - 7 | 1820 | 100 | |
AK131539.1 | - | 14 - 15 | 3402 | 100 | |
AK225786.1 | - | 13 - 14 | 3362 | 100 | |
BG742978.1 | - | 2 - 3 | 199 | 100 | |
L13738.2 | - | 14 - 15 | 3725 | 100 |