The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005858.3 | NP_005849.1 | 1 - 2 | 87 | 96 |
GenBank mRNA | BC037270.2 | - | 1 - 2 | 77 | 96 |
DR005936.1 | - | 1 - 2 | 90 | 96 | |
Y11997.1 | - | 1 - 2 | 78 | 96 |