The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258460.1 | NP_001245389.1 | 4 - 5 | 481 | 122 |
NM_006213.4 | NP_006204.1 | 4 - 5 | 508 | 122 | |
XM_017012324.1 | XP_016867813.1 | 4 - 5 | 508 | 122 | |
GenBank mRNA | AK300928.1 | - | 4 - 5 | 433 | 122 |
AK312851.1 | - | 4 - 5 | 508 | 122 | |
BC051327.1 | - | 4 - 5 | 446 | 122 | |
BC069679.1 | - | 4 - 5 | 424 | 122 | |
BC069738.1 | - | 4 - 5 | 425 | 122 | |
BC069754.1 | - | 4 - 5 | 425 | 122 | |
BC074753.2 | - | 3 - 4 | 352 | 122 | |
DB171823.1 | - | 4 - 5 | 508 | 122 | |
X80590.1 | - | 4 - 5 | 432 | 122 |