The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256045.1 | NP_001242974.1 | 2 - 3 | 657 | 85 |
NM_006219.2 | NP_006210.1 | 11 - 12 | 1791 | 85 | |
XM_005247530.2 | XP_005247587.1 | 13 - 14 | 2140 | 85 | |
XM_006713659.3 | XP_006713722.1 | 12 - 13 | 2011 | 85 | |
XM_011512895.2 | XP_011511197.1 | 14 - 15 | 2136 | 85 | |
XM_011512896.1 | XP_011511198.1 | 9 - 10 | 1405 | 85 | |
XM_017006619.1 | XP_016862108.1 | 12 - 13 | 1814 | 85 | |
XM_017006620.1 | XP_016862109.1 | 11 - 12 | 1561 | 85 | |
XM_017006621.1 | XP_016862110.1 | 10 - 11 | 1510 | 85 | |
GenBank mRNA | AK293754.1 | - | 2 - 3 | 657 | 85 |
AK302937.1 | - | 4 - 5 | 670 | 85 | |
BC114432.1 | - | 11 - 12 | 1779 | 85 | |
CR749357.1 | - | 4 - 5 | 361 | 85 | |
S67334.1 | - | 11 - 12 | 1775 | 85 |