Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12648096-12648590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAN2B1
Entrez Gene ID
4125
Gene Name
mannosidase alpha class 2B member 1
Gene Aliases
LAMAN, MANB
UniGene
Hs.356769
Gene Chromosome Location
Chr.19: 12646508-12666777 on Build GRCh38
Transcripts
NM_000528.3 NM_001173498.1 XM_005259913.1 XM_017026818.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4074379 c.2504C>T p.A835V Chr.19: 12648332-12648332 on Build GRCh38
5388670 c.2500G>A p.G834R Chr.19: 12648336-12648336 on Build GRCh38
4700121 c.2556C>T p.A852A Chr.19: 12648280-12648280 on Build GRCh38
7302979 c.2513G>A p.R838Q Chr.19: 12648323-12648323 on Build GRCh38
3528785 c.2521C>T p.H841Y Chr.19: 12648315-12648315 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760097655 Chr.19: 12648330-12648330 on Build GRCh38
rs1051322269 Chr.19: 12648234-12648234 on Build GRCh38
rs755344489 Chr.19: 12648232-12648232 on Build GRCh38
rs751431290 Chr.19: 12648428-12648429 on Build GRCh38
rs769427198 Chr.19: 12648405-12648405 on Build GRCh38
rs192419876 Chr.19: 12648505-12648505 on Build GRCh38
rs768244588 Chr.19: 12648290-12648290 on Build GRCh38
rs189469608 Chr.19: 12648425-12648425 on Build GRCh38
rs199967717 Chr.19: 12648338-12648338 on Build GRCh38
rs931486929 Chr.19: 12648532-12648532 on Build GRCh38
rs144244650 Chr.19: 12648342-12648342 on Build GRCh38
rs776299948 Chr.19: 12648305-12648305 on Build GRCh38
rs775212002 Chr.19: 12648178-12648178 on Build GRCh38
rs776013861 Chr.19: 12648412-12648412 on Build GRCh38
rs774405585 Chr.19: 12648156-12648156 on Build GRCh38
rs1046211352 Chr.19: 12648184-12648184 on Build GRCh38
rs545541738 Chr.19: 12648217-12648217 on Build GRCh38
rs747744816 Chr.19: 12648391-12648391 on Build GRCh38
rs755996633 Chr.19: 12648442-12648443 on Build GRCh38
rs1016249959 Chr.19: 12648522-12648522 on Build GRCh38
rs530923717 Chr.19: 12648460-12648460 on Build GRCh38
rs369174673 Chr.19: 12648374-12648374 on Build GRCh38
rs778022209 Chr.19: 12648256-12648256 on Build GRCh38
rs540034802 Chr.19: 12648352-12648352 on Build GRCh38
rs376734029 Chr.19: 12648428-12648428 on Build GRCh38
rs550301551 Chr.19: 12648150-12648150 on Build GRCh38
rs933863283 Chr.19: 12648489-12648489 on Build GRCh38
rs979496069 Chr.19: 12648213-12648213 on Build GRCh38
rs758456823 Chr.19: 12648426-12648426 on Build GRCh38
rs763600117 Chr.19: 12648277-12648277 on Build GRCh38
rs561895292 Chr.19: 12648370-12648370 on Build GRCh38
rs1046879893 Chr.19: 12648502-12648502 on Build GRCh38
rs958869615 Chr.19: 12648361-12648361 on Build GRCh38
rs764660688 Chr.19: 12648319-12648319 on Build GRCh38
rs956839427 Chr.19: 12648437-12648437 on Build GRCh38
rs775204651 Chr.19: 12648278-12648278 on Build GRCh38
rs528323073 Chr.19: 12648288-12648288 on Build GRCh38
rs570839557 Chr.19: 12648509-12648509 on Build GRCh38
rs1039625598 Chr.19: 12648470-12648470 on Build GRCh38
rs556554403 Chr.19: 12648181-12648181 on Build GRCh38
rs749425508 Chr.19: 12648260-12648260 on Build GRCh38
rs752079394 Chr.19: 12648321-12648321 on Build GRCh38
rs781357715 Chr.19: 12648241-12648241 on Build GRCh38
rs371508056 Chr.19: 12648436-12648436 on Build GRCh38
rs760186947 Chr.19: 12648188-12648188 on Build GRCh38
rs886750475 Chr.19: 12648340-12648340 on Build GRCh38
rs767484276 Chr.19: 12648435-12648436 on Build GRCh38
rs1012498827 Chr.19: 12648552-12648552 on Build GRCh38
rs759423485 Chr.19: 12648162-12648162 on Build GRCh38
rs535248547 Chr.19: 12648533-12648533 on Build GRCh38
rs763786776 Chr.19: 12648199-12648199 on Build GRCh38
rs368834616 Chr.19: 12648214-12648214 on Build GRCh38
rs773223523 Chr.19: 12648534-12648534 on Build GRCh38
rs750339878 Chr.19: 12648436-12648437 on Build GRCh38
rs759144415 Chr.19: 12648553-12648553 on Build GRCh38
rs745928175 Chr.19: 12648360-12648360 on Build GRCh38
rs200695002 Chr.19: 12648407-12648407 on Build GRCh38
rs761344747 Chr.19: 12648313-12648313 on Build GRCh38
rs1046850193 Chr.19: 12648339-12648339 on Build GRCh38
rs140281123 Chr.19: 12648333-12648333 on Build GRCh38
rs747937275 Chr.19: 12648445-12648445 on Build GRCh38
rs991714132 Chr.19: 12648143-12648143 on Build GRCh38
rs746898354 Chr.19: 12648433-12648433 on Build GRCh38
rs772778684 Chr.19: 12648406-12648406 on Build GRCh38
rs781026700 Chr.19: 12648438-12648438 on Build GRCh38
rs372896479 Chr.19: 12648220-12648220 on Build GRCh38
rs756475764 Chr.19: 12648334-12648334 on Build GRCh38
rs1030405860 Chr.19: 12648327-12648327 on Build GRCh38
rs752022224 Chr.19: 12648221-12648221 on Build GRCh38
rs935084853 Chr.19: 12648443-12648443 on Build GRCh38
rs1040271670 Chr.19: 12648292-12648292 on Build GRCh38
rs771953225 Chr.19: 12648174-12648174 on Build GRCh38
rs914908585 Chr.19: 12648458-12648458 on Build GRCh38
rs970921118 Chr.19: 12648386-12648386 on Build GRCh38
rs886054229 Chr.19: 12648281-12648281 on Build GRCh38
rs550855892 Chr.19: 12648424-12648424 on Build GRCh38
rs866046032 Chr.19: 12648317-12648317 on Build GRCh38
rs752103191 Chr.19: 12648236-12648236 on Build GRCh38
rs920051618 Chr.19: 12648514-12648514 on Build GRCh38
rs1021381108 Chr.19: 12648408-12648408 on Build GRCh38
rs764448970 Chr.19: 12648420-12648420 on Build GRCh38
rs756359957 Chr.19: 12648255-12648255 on Build GRCh38
rs991095656 Chr.19: 12648346-12648346 on Build GRCh38
rs746805353 Chr.19: 12648372-12648372 on Build GRCh38
rs11554969 Chr.19: 12648222-12648222 on Build GRCh38
rs758333025 Chr.19: 12648365-12648365 on Build GRCh38
rs757416569 Chr.19: 12648351-12648351 on Build GRCh38
rs753095085 Chr.19: 12648249-12648249 on Build GRCh38
rs760974907 Chr.19: 12648418-12648418 on Build GRCh38
rs10410289 Chr.19: 12648277-12648277 on Build GRCh38
rs757044622 Chr.19: 12648429-12648430 on Build GRCh38

View Full Product Details