The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005178.4 | NP_005169.2 | 2 - 3 | 480 | 60 |
XM_011527197.2 | XP_011525499.2 | 4 - 5 | 890 | 60 | |
XM_011527198.2 | XP_011525500.2 | 5 - 6 | 1272 | 60 | |
XM_011527200.2 | XP_011525502.1 | 2 - 3 | 956 | 60 | |
XM_017027109.1 | XP_016882598.1 | 2 - 3 | 1914 | 60 | |
XM_017027110.1 | XP_016882599.1 | 2 - 3 | 1920 | 60 | |
GenBank mRNA | AK296180.1 | - | 2 - 3 | 578 | 60 |
BC064993.1 | - | 2 - 3 | 478 | 60 | |
BU501187.1 | - | 2 - 3 | 480 | 60 | |
M31732.1 | - | 2 - 3 | 427 | 60 |