The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006099.3 | NP_006090.2 | 7 - 8 | 1001 | 102 |
GenBank mRNA | AB021868.1 | - | 7 - 8 | 908 | 102 |
AK027557.1 | - | 7 - 8 | 1000 | 102 | |
BC001154.1 | - | 7 - 8 | 946 | 102 | |
BC030556.1 | - | 7 - 8 | 957 | 102 | |
BT007036.1 | - | 6 - 7 | 883 | 102 | |
CR457090.1 | - | 6 - 7 | 883 | 102 |