Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDP
Entrez Gene ID
4693
Gene Name
NDP, norrin cystine knot growth factor
Gene Aliases
EVR2, FEVR, ND
Chromosome Location
Chr.X: 43948776 - 43973675 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000266.3 NP_000257.1 1 - 2 376 110
GenBank mRNA X65724.1 - 1 - 2 213 110
X65882.1 - 1 - 2 205 110

View Full Product Details