The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000639.2 | NP_000630.1 | 3 - 4 | 649 | 56 |
NM_001302746.1 | NP_001289675.1 | 2 - 3 | 603 | 56 | |
GenBank mRNA | AF288573.1 | - | 2 - 3 | 467 | 56 |
AK314874.1 | - | 3 - 4 | 613 | 56 | |
AY225406.1 | - | 3 - 4 | 455 | 56 | |
AY858799.1 | - | 3 - 4 | 455 | 56 | |
BC017502.1 | - | 3 - 4 | 542 | 56 | |
D38122.1 | - | 3 - 4 | 643 | 56 | |
KJ948655.1 | - | 3 - 4 | 612 | 56 | |
U08137.1 | - | 3 - 4 | 540 | 56 | |
U11821.1 | - | 3 - 4 | 519 | 56 | |
X89102.1 | - | 3 - 4 | 612 | 56 |