Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHRNB2
Entrez Gene ID
1141
Gene Name
cholinergic receptor nicotinic beta 2 subunit
Gene Aliases
EFNL3, nAChRB2
Chromosome Location
Chr.1: 154567781 - 154580026 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000748.2 NP_000739.1 4 - 5 635 62
GenBank mRNA AK313470.1 - 4 - 5 404 62
BC075040.2 - 4 - 5 400 62
BC075041.2 - 4 - 5 400 62
U62437.1 - 4 - 5 635 62
X53179.1 - 4 - 5 537 62
Y08415.1 - 4 - 5 371 62

View Full Product Details