Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BMP4
Entrez Gene ID
652
Gene Name
bone morphogenetic protein 4
Gene Aliases
BMP2B, BMP2B1, MCOPS6, OFC11, ZYME
Chromosome Location
Chr.14: 53949736 - 53956862 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202.4 NP_001193.2 1 - 2 312 60
XM_017021604.1 XP_016877093.1 1 - 2 272 60
GenBank mRNA BQ083000.1 - 1 - 2 312 60
M22490.1 - 1 - 2 261 60

View Full Product Details