Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRF
Entrez Gene ID
6722
Gene Name
serum response factor
Gene Aliases
MCM1
Chromosome Location
Chr.6: 43171295 - 43181506 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003131.3 NP_003122.1 1 - 2 878 67
GenBank mRNA AB209128.1 - 1 - 2 626 67
BC048211.1 - 1 - 2 714 67
BC052572.1 - 1 - 2 878 67
J03161.1 - 1 - 2 874 67

View Full Product Details