The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286500.1 | NP_001273429.1 | 1 - 2 | 130 | 72 |
NM_003905.3 | NP_003896.1 | 1 - 2 | 130 | 72 | |
XM_005256215.1 | XP_005256272.1 | 1 - 2 | 131 | 72 | |
GenBank mRNA | AK097680.1 | - | 1 - 2 | 130 | 72 |
AK312784.1 | - | 1 - 2 | 130 | 72 | |
AL136798.1 | - | 1 - 2 | 99 | 72 | |
AY197612.1 | - | 1 - 2 | 68 | 72 | |
BC000480.2 | - | 1 - 2 | 61 | 72 | |
BC013301.2 | - | 1 - 2 | 61 | 72 | |
BM785899.1 | - | 1 - 2 | 130 | 72 | |
U50939.1 | - | 1 - 2 | 126 | 72 |