Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FGF14
Entrez Gene ID
2259
Gene Name
fibroblast growth factor 14
Gene Aliases
FGF-14, FHF-4, FHF4, SCA27
Chromosome Location
Chr.13: 101720855 - 102402428 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004115.3 NP_004106.1 1 - 2 196 82
GenBank mRNA U66200.1 - 1 - 2 196 82

View Full Product Details