The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004291.3 | NP_004282.1 | 1 - 2 | 295 | 77 |
GenBank mRNA | BC029882.1 | - | 1 - 2 | 291 | 77 |
CR542216.1 | - | 1 - 2 | 164 | 77 | |
U16826.1 | - | 1 - 2 | 183 | 77 |