The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303422.1 | NP_001290351.1 | 5 - 6 | 727 | 125 |
NM_004490.2 | NP_004481.2 | 6 - 7 | 1356 | 125 | |
XM_011511022.2 | XP_011509324.1 | 6 - 7 | 988 | 125 | |
XM_017003899.1 | XP_016859388.1 | 6 - 7 | 793 | 125 | |
GenBank mRNA | AK301961.1 | - | 5 - 6 | 672 | 125 |
AK302129.1 | - | 5 - 6 | 684 | 125 | |
AK307744.1 | - | 6 - 7 | 868 | 125 | |
AK313153.1 | - | 6 - 7 | 985 | 125 | |
BC053559.1 | - | 6 - 7 | 826 | 125 | |
L76687.1 | - | 6 - 7 | 1355 | 125 |