The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004841.3 | NP_004832.1 | 3 - 4 | 358 | 70 |
NM_170692.2 | NP_733793.2 | 5 - 6 | 1441 | 70 | |
XM_005245622.3 | XP_005245679.1 | 3 - 4 | 1014 | 70 | |
XM_011510166.2 | XP_011508468.1 | 5 - 6 | 1441 | 70 | |
XM_011510167.2 | XP_011508469.1 | 5 - 6 | 1441 | 70 | |
XM_017002849.1 | XP_016858338.1 | 5 - 6 | 4047 | 70 | |
XM_017002850.1 | XP_016858339.1 | 5 - 6 | 4046 | 70 | |
XM_017002851.1 | XP_016858340.1 | 5 - 6 | 4046 | 70 | |
XM_017002852.1 | XP_016858341.1 | 5 - 6 | 4045 | 70 | |
XM_017002853.1 | XP_016858342.1 | 5 - 6 | 1485 | 70 | |
XM_017002854.1 | XP_016858343.1 | 3 - 4 | 1017 | 70 | |
XM_017002855.1 | XP_016858344.1 | 3 - 4 | 1016 | 70 | |
GenBank mRNA | AB209789.1 | - | 5 - 6 | 1349 | 70 |
AF047711.1 | - | 3 - 4 | 358 | 70 | |
AK075169.1 | - | 5 - 6 | 1441 | 70 | |
BC041045.1 | - | - | 994 | 70 | |
BC063442.1 | - | 5 - 6 | 1001 | 70 | |
BC078167.1 | - | 5 - 6 | 1063 | 70 | |
BC110611.2 | - | 5 - 6 | 673 | 70 |