The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017915.2 | NP_001017915.1 | 22 - 23 | 2958 | 55 |
NM_005541.4 | NP_005532.2 | 22 - 23 | 2955 | 55 | |
XM_017004004.1 | XP_016859493.1 | 22 - 23 | 2534 | 55 | |
GenBank mRNA | BC099920.1 | - | 22 - 23 | 2566 | 55 |
BC113580.1 | - | 22 - 23 | 2503 | 55 | |
BC113582.1 | - | 22 - 23 | 2503 | 55 | |
U50040.1 | - | 17 - 18 | 1823 | 55 | |
U50041.1 | - | 22 - 23 | 2567 | 55 | |
U53470.1 | - | 22 - 23 | 2576 | 55 | |
U57650.1 | - | 22 - 23 | 2955 | 55 | |
U84400.1 | - | 22 - 23 | 2527 | 55 | |
X98429.1 | - | 22 - 23 | 2583 | 55 |