The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006206.4 | NP_006197.1 | 1 - 2 | 319 | 92 |
XM_017008282.1 | XP_016863771.1 | 1 - 2 | 248 | 92 | |
GenBank mRNA | AK308353.1 | - | 1 - 2 | 149 | 92 |
AK311006.1 | - | 1 - 2 | 124 | 92 | |
AK316578.1 | - | 1 - 2 | 121 | 92 | |
BC015186.1 | - | 1 - 2 | 136 | 92 | |
BC063414.1 | - | 1 - 2 | 103 | 92 | |
DA678599.1 | - | 1 - 2 | 319 | 92 | |
M21574.1 | - | 1 - 2 | 127 | 92 | |
M22734.1 | - | - | 116 | 92 |