The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190995.1 | NP_001177924.1 | 16 - 17 | 1977 | 71 |
NM_006390.3 | NP_006381.2 | 20 - 21 | 2615 | 71 | |
XM_017018691.1 | XP_016874180.1 | 20 - 21 | 2512 | 71 | |
XM_017018692.1 | XP_016874181.1 | 19 - 20 | 2172 | 71 | |
GenBank mRNA | AB209602.1 | - | 7 - 8 | 695 | 71 |
AK302260.1 | - | 16 - 17 | 1943 | 71 | |
AU132276.3 | - | 5 - 6 | 492 | 71 | |
U77494.1 | - | 20 - 21 | 2437 | 71 |