The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018659.2 | NP_061129.1 | 1 - 2 | 182 | 70 |
GenBank mRNA | AF193766.1 | - | 1 - 2 | 185 | 70 |
AF274956.1 | - | 1 - 2 | 86 | 70 | |
AY359101.1 | - | 1 - 2 | 183 | 70 | |
BC031391.1 | - | 1 - 2 | 158 | 70 |