The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040875.2 | NP_001035808.1 | 2 - 3 | 260 | 96 |
NM_018972.2 | NP_061845.2 | 2 - 3 | 390 | 96 | |
XM_017013585.1 | XP_016869074.1 | 2 - 3 | 1522 | 96 | |
XM_017013586.1 | XP_016869075.1 | 2 - 3 | 1522 | 96 | |
GenBank mRNA | AB551556.1 | - | 2 - 3 | 389 | 96 |
AB551557.1 | - | 2 - 3 | 389 | 96 | |
AB551558.1 | - | 2 - 3 | 387 | 96 | |
AK292572.1 | - | 2 - 3 | 329 | 96 | |
BC024939.1 | - | 2 - 3 | 225 | 96 | |
BC035450.1 | - | 2 - 3 | 237 | 96 | |
DB505741.1 | - | 2 - 3 | 390 | 96 | |
Y17849.1 | - | 2 - 3 | 318 | 96 |