The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253723.1 | NP_001240652.1 | 3 - 4 | 499 | 89 |
NM_001253724.1 | NP_001240653.1 | 3 - 4 | 502 | 89 | |
NM_020134.3 | NP_064519.2 | 3 - 4 | 582 | 89 | |
GenBank mRNA | AF157634.1 | - | 2 - 3 | 424 | 89 |
AF264015.1 | - | 3 - 4 | 586 | 89 | |
AJ251275.1 | - | 2 - 3 | 424 | 89 | |
AL713706.1 | - | 1 - 2 | 124 | 89 | |
BC002874.2 | - | 3 - 4 | 499 | 89 | |
BT006871.1 | - | 2 - 3 | 424 | 89 | |
DA410946.1 | - | 3 - 4 | 502 | 89 |