The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001695.4 | NP_001686.1 | 1 - 2 | 205 | 88 |
GenBank mRNA | AK000064.1 | - | 1 - 2 | 133 | 88 |
AL707882.1 | - | 1 - 2 | 210 | 88 | |
AU137773.2 | - | 1 - 2 | 149 | 88 | |
AV649881.2 | - | 1 - 2 | 101 | 88 | |
BC010960.1 | - | 1 - 2 | 131 | 88 | |
X69151.1 | - | 1 - 2 | 126 | 88 |