Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSSC1
Entrez Gene ID
7260
Gene Name
tumor suppressing subtransferable candidate 1
Gene Aliases
-
Chromosome Location
Chr.2: 3188968 - 3377882 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003310.2 NP_003301.1 1 - 2 237 71
XM_011510384.2 XP_011508686.1 1 - 2 185 71
XM_011510387.2 XP_011508689.1 1 - 2 185 71
GenBank mRNA AF019952.1 - 1 - 2 195 71
AK026522.1 - 1 - 2 178 71
AK222652.1 - 1 - 2 173 71
BC002485.2 - 1 - 2 145 71
DA349228.1 - 1 - 2 237 71

View Full Product Details