Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDLIM4
Entrez Gene ID
8572
Gene Name
PDZ and LIM domain 4
Gene Aliases
RIL
Chromosome Location
Chr.5: 132257658 - 132273454 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001131027.1 NP_001124499.1 2 - 3 323 80
NM_003687.3 NP_003678.2 2 - 3 323 80
GenBank mRNA AF153882.1 - 2 - 3 80
AF154335.1 - 2 - 3 80
AK225407.1 - 2 - 3 285 80
AK313508.1 - 2 - 3 80
BC003096.1 - 2 - 3 80
BC016765.1 - 2 - 3 80
BT007019.1 - 2 - 3 80
DN997097.1 - 2 - 3 342 80
KU178433.1 - 2 - 3 246 80
KU178434.1 - 2 - 3 246 80
X93510.1 - 2 - 3 80

View Full Product Details