Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RARRES3
Entrez Gene ID
5920
Gene Name
retinoic acid receptor responder 3
Gene Aliases
HRASLS4, HRSL4, PLA1/2-3, RIG1, TIG3
Chromosome Location
Chr.11: 63536801 - 63546458 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004585.4 NP_004576.2 1 - 2 84 61
GenBank mRNA AB030815.1 - 1 - 2 40 61
AF060228.1 - 1 - 2 45 61
AF092922.1 - 1 - 2 77 61
AK293357.1 - 1 - 2 69 61
AK312110.1 - 1 - 2 69 61
BC009678.1 - 1 - 2 64 61
DA973023.1 - 1 - 2 86 61

View Full Product Details